Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291780.1 | NP_001278709.1 | 1 | 287 | ||
NM_017495.5 | NP_059965.2 | 1 | 287 | |||
NM_183425.2 | NP_906270.1 | 1 | 287 | |||
XM_005260446.3 | XP_005260503.1 | 1 | 225 | |||
XM_011528885.2 | XP_011527187.1 | 1 | 219 | |||
GenBank mRNA | AF432218.1 | - | 1 | 283 | ||
BC018711.1 | - | 1 | 39 | 4852523 | ||
BM549757.1 | - | 282 | ||||
DN998561.1 | - | 1 | 272 | |||
X75314.1 | - | 1 | 74 |