Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144703.2 | NP_653304.2 | 6 | 964 | ||
XM_005260302.3 | XP_005260359.1 | 7 | 1076 | |||
XM_011528605.2 | XP_011526907.1 | 8 | 1268 | |||
XM_011528606.2 | XP_011526908.1 | 8 | 1186 | |||
XM_011528607.2 | XP_011526909.1 | 7 | 1194 | |||
XM_011528608.2 | XP_011526910.1 | 7 | 1157 | |||
XM_011528609.2 | XP_011526911.1 | 6 | 1038 | |||
XM_011528611.2 | XP_011526913.1 | 5 | 882 | |||
XM_011528612.2 | XP_011526914.1 | 8 | 938 | |||
XM_011528613.2 | XP_011526915.1 | 7 | 798 | |||
XM_011528614.2 | XP_011526916.1 | 7 | 1011 | |||
XM_011528615.2 | XP_011526917.1 | 4 | 495 | |||
XM_017027688.1 | XP_016883177.1 | 5 | 871 | |||
XM_017027689.1 | XP_016883178.1 | 5 | 790 | |||
XM_017027690.1 | XP_016883179.1 | 5 | 612 | |||
GenBank mRNA | AK058202.1 | - | 8 | 943 | ||
AK298748.1 | - | 6 | 673 | |||
BC035702.1 | - | 6 | 1029 | |||
BX370449.2 | - | 2 | 164 |