Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152372.3 | NP_689585.3 | 32 | 3909 | ||
GenBank mRNA | AK093280.1 | - | 5 | 795 | ||
AK128233.1 | - | 24 | 2969 | |||
BC014671.1 | - | 6 | 429 | |||
BX537911.1 | - | 31 | 3633 | |||
BX647453.1 | - | 32 | 3870 | |||
BX647644.1 | - | 32 | 3870 |