Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307940.1 | NP_001294869.1 | 2 | 238 | ||
NM_024921.3 | NP_079197.3 | 2 | 238 | |||
XM_005262203.3 | XP_005262260.1 | 2 | 196 | |||
GenBank mRNA | AF309774.1 | - | 2 | 238 | ||
AK025039.1 | - | 2 | 187 | |||
AK290360.1 | - | 2 | 187 | |||
AK314295.1 | - | 2 | 157 | |||
BC017500.1 | - | 2 | 212 | 4905554 |