Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016024.2 | NP_057108.2 | 5 | 499 | ||
GenBank mRNA | AF078865.1 | - | 4 | 435 | ||
AF151837.1 | - | 4 | 435 | |||
AK292855.1 | - | 5 | 481 | |||
AL050405.1 | - | 5 | 474 | |||
BC003676.1 | - | 5 | 499 | |||
BC033750.1 | - | 5 | 467 | 5210341 | ||
BC040433.1 | - | 5 | 450 | |||
BC105582.1 | - | 5 | 486 |