Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172702.1 | NP_001166173.1 | 1, 2 | 273 | ||
NM_153811.2 | NP_722518.2 | 1, 2 | 273 | |||
XM_006720049.3 | XP_006720112.1 | 1, 2 | 225 | |||
XM_011536469.2 | XP_011534771.1 | 1, 2 | 225 | |||
XM_017021020.1 | XP_016876509.1 | 1, 2 | 225 | |||
XM_017021021.1 | XP_016876510.1 | 1, 2 | 225 | |||
XM_017021023.1 | XP_016876512.1 | 1, 2 | 225 | |||
XM_017021024.1 | XP_016876513.1 | 1, 2 | 225 | |||
GenBank mRNA | AF543422.1 | - | 1, 2 | 225 | ||
BC110378.1 | - | 1, 2 | 134 | 5737780 | ||
BP352279.1 | - | 1, 2 | 150 | |||
BX248072.1 | - | 1, 2 | 94 | |||
BX648332.1 | - | 1, 2 | 147 | |||
DR002669.1 | - | 1, 2 | 280 |