Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHERP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006387.5 | NP_006378.3 | 8 | 963 | ||
GenBank mRNA | AF536542.1 | - | 8 | 930 | ||
BC001552.2 | - | 8 | 921 | |||
BC021294.2 | - | 8 | 948 | 4053575 | ||
U94836.2 | - | 7 | 865 | |||
Y08265.1 | - | 8 | 920 |