Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144992.4 | NP_659429.4 | 22 | 3170 | ||
XM_005263897.2 | XP_005263954.1 | 22 | 3135 | |||
XM_006712357.2 | XP_006712420.1 | 21 | 2884 | |||
XM_006712359.3 | XP_006712422.1 | 13 | 1564 | |||
XM_006712360.3 | XP_006712423.1 | 10 | 1211 | |||
XM_011510770.1 | XP_011509072.1 | 21 | 3084 | |||
XM_011510771.2 | XP_011509073.1 | 21 | 3009 | |||
XM_011510772.1 | XP_011509074.1 | 22 | 3135 | |||
XM_017003560.1 | XP_016859049.1 | 16 | 2001 | |||
XM_017003561.1 | XP_016859050.1 | 16 | 2007 | |||
XM_017003562.1 | XP_016859051.1 | 16 | 2057 | |||
XM_017003563.1 | XP_016859052.1 | 16 | 2007 | |||
GenBank mRNA | AK090786.1 | - | 9 | 1006 | ||
AK093084.1 | - | 10 | 1137 | |||
AK098841.1 | - | 4 | 409 | |||
AK295743.1 | - | 16 | 2065 | |||
AL832635.2 | - | 23 | 3113 | |||
AL832761.1 | - | 23 | 3341 | |||
AL834173.1 | - | 13 | 1569 | |||
AL834385.1 | - | 12 | 1433 | |||
BC022028.2 | - | 4 | 400 | |||
BC150653.1 | - | 22 | 3110 | 9057024 |