Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNNM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020348.2 | NP_065081.2 | 9, 10 | 2971 | ||
XM_011539629.1 | XP_011537931.1 | 10, 11 | 3034 | |||
GenBank mRNA | AF169226.1 | - | 9, 10 | 1794 | ||
AK056194.1 | - | 8, 9 | 1074 | |||
AK296545.1 | - | 10, 11 | 2202 | |||
AK310486.1 | - | 8, 9 | 1226 | |||
AL137536.1 | - | 2, 3 | 342 | |||
BC098103.1 | - | 9, 10 | 1745 | 40006972 | ||
BC098279.1 | - | 9, 10 | 1745 | 40006977 | ||
BC098307.1 | - | 9, 10 | 1745 | 40006978 | ||
JF267782.1 | - | 7, 8 | 901 |