Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021195.4 | NP_067018.2 | 2 | 992 | ||
GenBank mRNA | AK075329.1 | - | 2 | 989 | ||
AY358480.1 | - | 1 | 937 | |||
BC008934.2 | - | 1 | 966 | 2963008 |