Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRN3P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003370.2 | 12 | 1140 | |||
GenBank mRNA | AK126166.1 | - | 6 | 3218 | ||
AK291749.1 | - | 6 | 770 | |||
BC006441.1 | - | 6 | 2139 | |||
BC112115.1 | - | 6 | 1268 | |||
BC112117.1 | - | 6 | 1268 | |||
BC143685.1 | - | 6 | 1256 |