Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152479.5 | NP_689692.2 | 3 | 809 | ||
XM_017026368.1 | XP_016881857.1 | 3 | 1407 | |||
GenBank mRNA | AK054935.1 | - | 1 | 2291 | ||
AK289550.1 | - | 3 | 794 | |||
BC029539.1 | - | 3 | 801 | 5303555 |