Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161009.2 | NP_001154481.1 | 6 | 1864 | ||
NM_001161010.2 | NP_001154482.1 | 6 | 1792 | |||
NM_001161011.2 | NP_001154483.1 | 5 | 1792 | |||
NM_001161012.2 | NP_001154484.1 | 5 | 1687 | |||
NM_001161013.2 | NP_001154485.1 | 4 | 1615 | |||
NM_031905.4 | NP_114111.2 | 7 | 1969 | |||
XM_011516601.2 | XP_011514903.1 | 6 | 2042 | |||
XM_011516602.2 | XP_011514904.1 | 6 | 1403 | |||
XM_017012684.1 | XP_016868173.1 | 8 | 2150 | |||
XM_017012685.1 | XP_016868174.1 | 7 | 2078 | |||
XM_017012686.1 | XP_016868175.1 | 7 | 1973 | |||
XM_017012687.1 | XP_016868176.1 | 6 | 1382 | |||
GenBank mRNA | AK075500.1 | - | 7 | 1710 | ||
AK225082.1 | - | 6 | 1597 | |||
AY150851.1 | - | 5 | 1687 | |||
AY150852.1 | - | 6 | 1792 | |||
AY150853.1 | - | 6 | 1864 | |||
AY150854.1 | - | 7 | 1969 | |||
BQ440425.1 | - | 1 | 646 | |||
BU941174.1 | - | 1 | 201 |