Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111038.1 | NP_001104508.1 | 14 | 1603 | ||
NM_001287150.1 | NP_001274079.1 | 14 | 1603 | |||
NM_145032.3 | NP_659469.3 | 14 | 1780 | |||
NR_105043.1 | 15 | 1694 | ||||
XM_005250205.3 | XP_005250262.1 | 15 | 1686 | |||
XM_005250207.3 | XP_005250264.1 | 14 | 1587 | |||
XM_005250208.3 | XP_005250265.1 | 15 | 1686 | |||
XM_005250209.2 | XP_005250266.1 | 14 | 1584 | |||
XM_006715898.2 | XP_006715961.1 | 9 | 1031 | |||
XM_011515928.2 | XP_011514230.1 | 15 | 1686 | |||
XM_011515929.2 | XP_011514231.1 | 15 | 1686 | |||
XM_011515930.2 | XP_011514232.1 | 14 | 1650 | |||
XM_011515932.2 | XP_011514234.1 | 15 | 1686 | |||
XM_017011850.1 | XP_016867339.1 | 15 | 1630 | |||
XM_017011851.1 | XP_016867340.1 | 15 | 1686 | |||
XM_017011852.1 | XP_016867341.1 | 15 | 1836 | |||
XM_017011853.1 | XP_016867342.1 | 14 | 1636 | |||
GenBank mRNA | AK095387.1 | - | 9 | 1003 | ||
AK097537.1 | - | 14 | 1780 | |||
AL713709.1 | - | 5 | 502 | |||
AY359239.1 | - | 14 | 1487 | |||
BC020572.2 | - | 8 | 857 | |||
BC026121.1 | - | 15 | 1697 | 4826546 | ||
BC031285.1 | - | 14 | 1595 | 5298780 |