Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNMAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103149.1 | NP_001096619.1 | 2 | 992 | ||
NM_018215.3 | NP_060685.2 | 2 | 992 | |||
XM_011527067.2 | XP_011525369.1 | 2 | 1006 | |||
GenBank mRNA | AK001643.1 | - | 11 | |||
AK096326.1 | - | 2 | 552 | |||
AK290218.1 | - | 3 | 875 | |||
AL834344.1 | - | 2 | 836 | |||
BC026026.1 | - | 1 | 25 | |||
BC032508.1 | - | 2 | 970 | 5246377 | ||
BC051688.1 | - | 2 | 949 | 5295669 | ||
BX649091.1 | - | 1 | 87 |