Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033547.3 | NP_291025.3 | 23 | 3041 | ||
XM_011545352.2 | XP_011543654.1 | 24 | 3210 | |||
GenBank mRNA | AF172822.1 | - | 10 | 1427 | ||
AL834171.1 | - | 23 | 2985 | |||
BC006369.1 | - | 11 | 1410 | |||
BC008013.1 | - | 19 | 2494 | |||
BC009859.2 | - | 13 | 1700 | |||
BC015664.2 | - | 24 | 3351 | 4554197 | ||
BE727962.1 | - | 403 |