Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033547.3 | NP_291025.3 | 16 | 2021 | ||
XM_011545352.2 | XP_011543654.1 | 17 | 2190 | |||
XM_017018560.1 | XP_016874049.1 | 16 | 2054 | |||
GenBank mRNA | AF172822.1 | - | 3 | 407 | ||
AK290866.1 | - | 16 | 2017 | |||
AL834171.1 | - | 16 | 1964 | |||
BC006369.1 | - | 4 | 390 | |||
BC008013.1 | - | 12 | 1474 | |||
BC009859.2 | - | 6 | 680 | |||
BC015664.2 | - | 17 | 2331 | 4554197 |