Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OCLM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022375.3 | NP_071770.1 | 1 | 518 | ||
GenBank mRNA | AF142063.1 | - | 1 | 632 | ||
BC069096.1 | - | 1 | 535 | 7216910 | ||
BC093987.1 | - | 1 | 67 | 7939832 | ||
BC093989.1 | - | 1 | 67 | 7939834 |