Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144952.1 | NP_001138424.1 | 31 | 4439 | ||
XM_011524914.2 | XP_011523216.1 | 31 | 4823 | |||
XM_011524915.2 | XP_011523217.1 | 31 | 4825 | |||
GenBank mRNA | AB040947.1 | - | 24 | 3470 | ||
AL137357.1 | - | 9 | 1347 | |||
BC045763.1 | - | 13 | 1967 | |||
BC066363.1 | - | 23 | 3339 |