Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144952.1 | NP_001138424.1 | 35 | 4781 | ||
XM_011524914.2 | XP_011523216.1 | 34 | 5108 | |||
XM_011524915.2 | XP_011523217.1 | 35 | 5167 | |||
GenBank mRNA | AB040947.1 | - | 28 | 3812 | ||
AK001694.1 | - | 2 | 168 | |||
AL137357.1 | - | 13 | 1689 | |||
BC045763.1 | - | 16 | 2252 | |||
BC066363.1 | - | 27 | 3681 |