Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114978.1 | NP_001108450.1 | 10 | 1384 | ||
NM_001114979.1 | NP_001108451.1 | 10 | 1384 | |||
NM_001114980.1 | NP_001108452.1 | 8 | 1154 | |||
NM_001114981.1 | NP_001108453.1 | 8 | 1154 | |||
NM_001114982.1 | NP_001108454.1 | 8 | 1154 | |||
NM_003722.4 | NP_003713.3 | 10 | 1384 | |||
XM_005247843.3 | XP_005247900.1 | 10 | 1819 | |||
XM_005247844.4 | XP_005247901.1 | 10 | 1547 | |||
XM_011513251.2 | XP_011511553.1 | 11 | 1425 | |||
XM_011513252.2 | XP_011511554.1 | 10 | 1357 | |||
XM_017007387.1 | XP_016862876.1 | 8 | 1150 | |||
GenBank mRNA | AB010153.1 | - | 8 | 1071 | ||
AB016072.1 | - | 10 | 1322 | |||
AB016073.1 | - | 10 | 1322 | |||
AB042841.1 | - | 7 | 758 | |||
AF075428.1 | - | 9 | 1178 | |||
AF075429.1 | - | 8 | 1013 | |||
AF075430.1 | - | 9 | 1178 | |||
AF075431.1 | - | 8 | 1013 | |||
AF075432.1 | - | 9 | 1178 | |||
AF075433.1 | - | 8 | 1013 | |||
AF091627.1 | - | 8 | 1138 | |||
AF116771.1 | - | 10 | 1322 | |||
AK304127.1 | - | 8 | 1154 | |||
BC039815.1 | - | 10 | 1384 | 5552611 | ||
GQ202689.1 | - | 8 | 1152 | |||
GQ202690.1 | - | 8 | 1152 | |||
KP881229.1 | - | 8 | 1018 | |||
KP881230.1 | - | 8 | 1006 | |||
KP881231.1 | - | 7 | 751 | |||
KU178435.1 | - | 10 | 1295 | |||
KU178436.1 | - | 10 | 1283 | |||
Y16961.1 | - | 10 | 1322 |