Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RX5-TAX1BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037928.1 | 15 | 6058 | |||
GenBank mRNA | AB209623.1 | - | 15 | 5846 |
Gene Symbol: TAX1BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204698.1 | NP_001191627.1 | 3 | 1078 | ||
NM_014604.3 | NP_055419.1 | 4 | 1156 | |||
GenBank mRNA | AF028823.2 | - | 4 | 1159 | ||
AF234997.1 | - | 4 | 1050 | |||
AF277318.1 | - | 4 | 1029 | |||
AK001327.1 | - | 2 | 1770 | |||
BC023980.1 | - | 4 | 1059 | 4069168 | ||
U90913.1 | - | 4 | 1036 |