Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195797.1 | NP_001182726.1 | 1 | 174 | ||
NM_015364.4 | NP_056179.3 | 1 | 174 | |||
XM_011517508.2 | XP_011515810.1 | 1 | 107 | |||
XM_017013299.1 | XP_016868788.1 | 1 | 107 | |||
XM_017013300.1 | XP_016868789.1 | 1 | 228 | |||
GenBank mRNA | AB018549.1 | - | 1 | 185 | ||
AB446498.1 | - | 1 | 60 | |||
AF168121.1 | - | 1 | 81 | |||
BC020690.1 | - | 1 | 84 | 4767246 | ||
BG435728.1 | - | 1 | 176 |