Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIPAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020137.4 | NP_064522.3 | 4, 5 | 241 | ||
XM_011543935.1 | XP_011542237.1 | 4, 5 | 241 | |||
XM_011543936.1 | XP_011542238.1 | 4, 5 | 241 | |||
XM_011543937.1 | XP_011542239.1 | 4, 5 | 288 | |||
XM_017029656.1 | XP_016885145.1 | 4, 5 | 241 | |||
XM_017029657.1 | XP_016885146.1 | 4, 5 | 612 | |||
XM_017029658.1 | XP_016885147.1 | 4, 5 | 1551 | |||
XM_017029659.1 | XP_016885148.1 | 4, 5 | 288 | |||
XM_017029660.1 | XP_016885149.1 | 4, 5 | 642 | |||
XM_017029661.1 | XP_016885150.1 | 4, 5 | 288 | |||
GenBank mRNA | AB032993.1 | - | 4, 5 | 186 | ||
AK292158.1 | - | 4, 5 | 212 | |||
BC101544.1 | - | 4, 5 | 239 | 8069050 | ||
BC101546.1 | - | 4, 5 | 239 | 8069052 | ||
BX647801.1 | - | 4, 5 | 240 | |||
BX647804.1 | - | 4, 5 | 240 | |||
CD109978.1 | - | 268 | ||||
DB099470.1 | - | 4, 5 | 241 |