Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBXN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014607.3 | NP_055422.1 | 7 | 945 | ||
GenBank mRNA | AK026971.1 | - | 7 | 568 | ||
AK095402.1 | - | 3 | 333 | |||
AK292829.1 | - | 7 | 638 | |||
AK304096.1 | - | 7 | 655 | |||
BC035594.2 | - | 8 | 777 | 5580880 | ||
BX647216.1 | - | 5 | 1237 | |||
BX647662.1 | - | 7 | 737 | |||
D87684.1 | - | 7 | 698 | |||
EF199840.1 | - | 7 | 762 |