Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYFIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033028.1 | NP_001028200.1 | 8 | 1636 | ||
NM_001287810.2 | NP_001274739.1 | 24 | 2907 | |||
NM_001324119.1 | NP_001311048.1 | 23 | 2736 | |||
NM_001324120.1 | NP_001311049.1 | 23 | 2915 | |||
NM_001324122.1 | NP_001311051.1 | 24 | 2936 | |||
NM_001324123.1 | NP_001311052.1 | 23 | 2815 | |||
NM_001324124.1 | NP_001311053.1 | 22 | 2698 | |||
NM_001324125.1 | NP_001311054.1 | 20 | 2497 | |||
NM_001324126.1 | NP_001311055.1 | 22 | 2686 | |||
NM_014608.4 | NP_055423.1 | 23 | 2788 | |||
XM_011543873.2 | XP_011542175.1 | 23 | 3137 | |||
XM_011543874.1 | XP_011542176.1 | 23 | 3170 | |||
XM_011543876.2 | XP_011542178.2 | 23 | 2738 | |||
XM_017022023.1 | XP_016877512.1 | 23 | 3143 | |||
XM_017022024.1 | XP_016877513.1 | 23 | 3151 | |||
GenBank mRNA | AK125784.1 | - | 2 | 1774 | ||
AK125936.1 | - | 22 | 2841 | |||
AK127094.1 | - | 8 | 1636 | |||
AK291604.1 | - | 23 | 2715 | |||
AY763577.1 | - | 23 | 2752 | |||
AY763578.1 | - | 24 | 2818 | |||
AY763579.1 | - | 8 | 1636 | |||
AY763580.1 | - | 2 | 1774 | |||
BC001306.2 | - | 6 | 604 | |||
BC005097.1 | - | 23 | 2679 | 3163591 | ||
BC063869.1 | - | 8 | 1430 | |||
D38549.1 | - | 23 | 2682 | |||
KJ534819.1 | - | 22 | 2627 |