Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACSTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002353.2 | NP_002344.2 | 1 | 459 | ||
GenBank mRNA | BC009409.1 | - | 1 | 206 | 3611756 | |
BT007255.1 | - | 1 | 121 | |||
CR542260.1 | - | 1 | 121 | |||
X13425.1 | - | 1 | 191 | |||
X77753.1 | - | 1 | 736 |