Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014611.2 | NP_055426.1 | 40, 41 | 6150 | ||
XM_006715405.3 | XP_006715468.1 | 4, 5 | 1628 | |||
XM_011535635.2 | XP_011533937.1 | 40, 41 | 5955 | |||
XM_011535636.2 | XP_011533938.1 | 40, 41 | 6120 | |||
GenBank mRNA | AF503925.1 | - | 40, 41 | 6079 |