Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004019.1 | NP_001004019.1 | 18 | 3534 | ||
NM_001165035.1 | NP_001158507.1 | 18 | 3791 | |||
NM_001998.2 | NP_001989.2 | 17 | 3393 | |||
XM_006713026.2 | XP_006713089.1 | 16 | 3326 | |||
GenBank mRNA | AK129883.1 | - | 2878 | |||
AK300913.1 | - | 13 | 1544 | |||
AK304777.1 | - | 17 | 3493 | |||
AK304827.1 | - | 18 | 3791 | |||
AL050095.1 | - | 12 | 1445 | |||
AY130459.1 | - | 18 | 3478 | |||
BC051690.1 | - | 17 | 3356 | 5269126 | ||
X82494.1 | - | 17 | 3337 |