Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGDCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004884.3 | NP_004875.2 | 14 | 3101 | ||
XM_011522241.1 | XP_011520543.1 | 14 | 3040 | |||
XM_011522243.1 | XP_011520545.1 | 13 | 2627 | |||
XM_011522244.1 | XP_011520546.1 | 13 | 2616 | |||
GenBank mRNA | AF063936.1 | - | 6 | 1330 | ||
AK095529.1 | - | 14 | 2496 |