Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KIAA1217 SIRNA Assays ›
Gene Symbol:
KIAA1217
Gene Name:
KIAA1217
Gene Aliases:
SKT
Chromosome Location:
Chr.10: 23694746 - 24547848 on Build GRCh38
UniGene ID:
Hs.445885
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
217222
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KIAA1217
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098500.2
NP_001091970.1
15
3562
NM_001282767.1
NP_001269696.1
13
3291
NM_001282768.1
NP_001269697.1
13
3291
NM_001282769.1
NP_001269698.1
9
2374
NM_001282770.1
NP_001269699.1
9
2328
NM_001321681.1
NP_001308610.1
10
2479
NM_019590.4
NP_062536.2
14
3396
XM_005252516.3
XP_005252573.1
9
2302
XM_011519552.2
XP_011517854.2
15
3451
XM_011519555.2
XP_011517857.2
14
3369
XM_011519558.2
XP_011517860.2
15
3453
XM_011519559.2
XP_011517861.2
13
3038
XM_011519562.2
XP_011517864.2
15
3451
XM_011519564.2
XP_011517866.1
13
3018
XM_011519565.2
XP_011517867.2
14
3142
XM_011519566.2
XP_011517868.2
13
3232
XM_017016416.1
XP_016871905.1
15
3477
XM_017016417.1
XP_016871906.1
13
3291
XM_017016418.1
XP_016871907.1
13
4113
XM_017016419.1
XP_016871908.1
12
4008
XM_017016420.1
XP_016871909.1
13
2860
XM_017016421.1
XP_016871910.1
15
3070
XM_017016422.1
XP_016871911.1
12
2755
XM_017016423.1
XP_016871912.1
10
2407
XM_017016424.1
XP_016871913.1
10
2479
XM_017016425.1
XP_016871914.1
15
3453
XM_017016426.1
XP_016871915.1
15
3457
XM_017016427.1
XP_016871916.1
14
3118
XM_017016428.1
XP_016871917.1
12
2639
XM_017016429.1
XP_016871918.1
16
3611
GenBank mRNA
AB033043.2
-
15
3562
AB291616.1
-
12
2818
AK300422.1
-
9
2313
AK303905.1
-
9
2082
AL157473.1
-
2
222
AL833280.1
-
14
3024
BC018764.1
-
9
2142
BC098577.1
-
9
2282
BC136521.1
-
13
2909
9020511
BC144227.1
-
13
2909
9052747
BX640796.1
-
9
2303
BX648451.1
-
9
2303
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
embryonic skeletal system development
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-9xzrh:80/100.66.79.6:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline