Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024100.3 | NP_077005.2 | 10 | 1507 | ||
XM_017027027.1 | XP_016882516.1 | 12 | 2033 | |||
GenBank mRNA | AK131009.1 | - | 4 | 650 | ||
BC000040.2 | - | 10 | 1492 | 3507673 | ||
BC001648.2 | - | 10 | 1487 | 2819452 |