Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PHLDB1 SIRNA Assays ›
Gene Symbol:
PHLDB1
Gene Name:
pleckstrin homology like domain family B member 1
Gene Aliases:
LL5A
Chromosome Location:
Chr.11: 118606296 - 118658038 on Build GRCh38
UniGene ID:
Hs.504062
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
217230
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PHLDB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001144758.2
NP_001138230.1
12
2855
NM_001144759.2
NP_001138231.1
12
2855
NM_015157.3
NP_055972.1
13
3078
XM_005271468.3
XP_005271525.1
12
3624
XM_005271469.3
XP_005271526.1
12
3624
XM_005271470.3
XP_005271527.1
12
3624
XM_005271471.3
XP_005271528.1
12
3624
XM_005271472.3
XP_005271529.1
12
3624
XM_005271473.3
XP_005271530.1
12
3624
XM_005271474.4
XP_005271531.1
12
3624
XM_005277680.3
XP_005277737.1
13
3750
XM_006718796.3
XP_006718859.1
13
2939
XM_006718797.3
XP_006718860.1
13
3753
XM_006718798.3
XP_006718861.1
13
3753
XM_006718799.3
XP_006718862.1
13
3753
XM_006718800.3
XP_006718863.1
13
3753
XM_006718801.3
XP_006718864.1
13
3753
XM_006718802.3
XP_006718865.1
13
3753
XM_006718803.3
XP_006718866.1
13
3753
XM_011542703.2
XP_011541005.1
13
3753
XM_011542704.2
XP_011541006.1
13
3753
XM_011542705.2
XP_011541007.1
13
3753
XM_011542706.2
XP_011541008.1
13
3753
XM_011542707.2
XP_011541009.1
12
3624
XM_011542708.2
XP_011541010.1
13
2942
XM_011542709.2
XP_011541011.1
13
3252
XM_011542710.2
XP_011541012.1
13
3753
XM_011542711.2
XP_011541013.1
10
2659
XM_011542713.2
XP_011541015.1
10
2653
XM_011542715.2
XP_011541017.1
7
2131
XM_017017407.1
XP_016872896.1
12
3621
XM_017017408.1
XP_016872897.1
13
3753
XM_017017409.1
XP_016872898.1
13
3753
XM_017017410.1
XP_016872899.1
12
3624
GenBank mRNA
AB014538.2
-
12
2832
AB094090.1
-
13
3078
AB177859.1
-
7
1959
AK027510.1
-
4
484
AK074070.2
-
12
2804
AK074918.1
-
6
691
AK125783.1
-
14
2082
AL832373.1
-
12
2812
BC013031.2
-
6
841
BC098586.1
-
12
2832
30342198
BX648176.1
-
7
1007
CR749783.1
-
13
2990
Back To Top
More Information
Gene Ontology Categories:
Function(s)
regulation of gastrulation
regulation of epithelial to mesenchymal transition
regulation of microtubule cytoskeleton organization
positive regulation of basement membrane assembly involved in embryonic body morphogenesis
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-pg4v9:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline