Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014624.3 | NP_055439.1 | 3 | 589 | ||
XM_017002033.1 | XP_016857522.1 | 3 | 412 | |||
GenBank mRNA | BC001431.2 | - | 3 | 394 | 3138609 | |
BC009017.1 | - | 3 | 570 | 4157026 |