Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RILPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319243.1 | NP_001306172.1 | 8 | 2236 | ||
NM_001319244.1 | NP_001306173.1 | 7 | 1697 | |||
NM_001319302.1 | NP_001306231.1 | 8 | 2647 | |||
NM_178314.4 | NP_847884.2 | 7 | 2148 | |||
GenBank mRNA | AK096697.1 | - | 9 | 2078 | ||
BC080626.1 | - | 7 | 1697 | 6339887 | ||
BC089444.1 | - | 7 | 1950 | 30559974 |