Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178454.4 | NP_848549.3 | 9 | 1158 | ||
XM_005270469.2 | XP_005270526.1 | 10 | 1216 | |||
XM_005270470.1 | XP_005270527.1 | 10 | 1339 | |||
XM_006710361.1 | XP_006710424.1 | 10 | 1176 | |||
XM_006710362.1 | XP_006710425.1 | 10 | 1149 | |||
XM_011540707.1 | XP_011539009.1 | 10 | 1157 | |||
XM_011540708.1 | XP_011539010.1 | 10 | 1237 | |||
XM_017000320.1 | XP_016855809.1 | 9 | 1100 | |||
XM_017000321.1 | XP_016855810.1 | 9 | 959 | |||
GenBank mRNA | AK075350.1 | - | 5 | 604 | ||
AL833098.1 | - | 9 | 1526 | |||
AY336747.1 | - | 9 | 1036 | |||
AY358492.1 | - | 9 | 1021 | |||
BC047025.1 | - | 8 | 973 | 5491665 | ||
BC091509.1 | - | 10 | 1250 | 30513000 |