Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIFO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181643.5 | NP_857594.2 | 3 | 333 | ||
XM_005270472.1 | XP_005270529.1 | 3 | 442 | |||
GenBank mRNA | AK074433.1 | - | 3 | 268 | ||
BC029237.1 | - | 2 | 222 | |||
BC050319.1 | - | 3 | 297 | 4837146 | ||
BC068603.1 | - | 3 | 257 | |||
BC093891.1 | - | 3 | 320 | 7939736 | ||
BC101501.1 | - | 3 | 321 | 8069007 | ||
HM237139.1 | - | 3 | 320 |