Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000579.3 | NP_000570.1 | 3 | 1829 | ||
NM_001100168.1 | NP_001093638.1 | 3 | 1594 | |||
GenBank mRNA | AK292132.1 | - | 2 | 1526 | ||
U54994.1 | - | 2 | 1526 |