Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMOD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304418.1 | NP_001291347.1 | 3 | 1368 | ||
NM_198271.4 | NP_938012.2 | 2 | 1462 | |||
GenBank mRNA | AK057852.1 | - | 2 | 989 | ||
AK300096.1 | - | 3 | 1341 | |||
AL832033.1 | - | 2 | 1433 | |||
AL832709.1 | - | 2 | 1467 | |||
BC121019.2 | - | 2 | 1341 | 40117347 |