Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YJEFN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094976.1 | - | 1 | 1313 | ||
AK126916.1 | - | 3 | 2089 | |||
BC093948.1 | - | 1 | 280 | |||
BC112285.1 | - | 1 | 280 |