Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134374.1 | NP_001127846.1 | 4 | 1437 | ||
NM_198572.2 | NP_940974.2 | 5 | 1577 | |||
XM_011517021.1 | XP_011515323.1 | 6 | 1682 | |||
XM_011517022.2 | XP_011515324.1 | 6 | 1968 | |||
XM_011517023.2 | XP_011515325.1 | 6 | 2986 | |||
XM_011517024.2 | XP_011515326.1 | 7 | 2207 | |||
XM_011517025.2 | XP_011515327.1 | 6 | 2642 | |||
XM_017013399.1 | XP_016868888.1 | 7 | 1501 | |||
GenBank mRNA | AB092352.1 | - | 5 | 1565 | ||
AK301710.1 | - | 4 | 1425 | |||
BC050390.1 | - | 5 | 1567 | 5742710 | ||
BC053547.1 | - | 5 | 1552 | 5743874 |