Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286615.1 | NP_001273544.1 | 28 | 3644 | ||
NM_001286616.1 | NP_001273545.1 | 27 | 3216 | |||
NM_178826.3 | NP_849148.2 | 27 | 3539 | |||
XM_011537911.2 | XP_011536213.1 | 30 | 3639 | |||
XM_011537912.2 | XP_011536214.1 | 30 | 3643 | |||
XM_011537913.2 | XP_011536215.1 | 29 | 3594 | |||
XM_011537914.2 | XP_011536216.1 | 27 | 3150 | |||
XM_011537915.2 | XP_011536217.1 | 28 | 3462 | |||
XM_011537916.2 | XP_011536218.1 | 28 | 3303 | |||
XM_017018808.1 | XP_016874297.1 | 27 | 3154 | |||
XM_017018809.1 | XP_016874298.1 | 29 | 3730 | |||
XM_017018810.1 | XP_016874299.1 | 27 | 3396 | |||
XM_017018811.1 | XP_016874300.1 | 26 | 3347 | |||
XM_017018812.1 | XP_016874301.1 | 26 | 5365 | |||
XM_017018813.1 | XP_016874302.1 | 25 | 3215 | |||
XM_017018814.1 | XP_016874303.1 | 28 | 5604 | |||
XM_017018815.1 | XP_016874304.1 | 28 | 5608 | |||
XM_017018816.1 | XP_016874305.1 | 28 | 3687 | |||
XM_017018817.1 | XP_016874306.1 | 28 | 5643 | |||
GenBank mRNA | AK091540.1 | - | 18 | 2160 | ||
AK091591.1 | - | 27 | 3328 | |||
AK092596.1 | - | 15 | 1726 | |||
AK316105.1 | - | 28 | 3283 |