Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003125.2 | NP_003116.2 | 2 | 456 | ||
GenBank mRNA | BC056240.1 | - | 2 | 460 | ||
M19888.1 | - | 2 | 460 | |||
M21300.1 | - | 2 | 456 | |||
M21301.1 | - | 2 | 456 |