Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGC70870 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003682.1 | 1 | 1947 | |||
GenBank mRNA | AK092162.1 | - | 1 | 1401 | ||
BC063803.1 | - | 1 | 1612 | |||
BC106037.1 | - | 1 | 1936 |