Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTFR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145838.1 | NP_001139310.1 | 6 | 950 | ||
NM_014637.3 | NP_055452.3 | 8 | 1181 | |||
GenBank mRNA | AK298945.1 | - | 7 | 679 | ||
AK316202.1 | - | 7 | 721 | |||
BC036116.1 | - | 8 | 1095 | 5299040 | ||
BC043498.2 | - | 8 | 1080 | 6057524 | ||
BX537854.1 | - | 6 | 842 | |||
BX647507.1 | - | 8 | 1100 | |||
CR456910.1 | - | 6 | 912 | |||
D13634.1 | - | 7 | 929 |