Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1211L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207362.2 | NP_997245.2 | 5, 6 | 835 | ||
XM_005263923.3 | XP_005263980.1 | 5, 6 | 815 | |||
XM_005263924.3 | XP_005263981.1 | 5, 6 | 815 | |||
XM_005263926.3 | XP_005263983.1 | 5, 6 | 700 | |||
XM_011511093.1 | XP_011509395.1 | 7, 8 | 974 | |||
XM_011511094.1 | XP_011509396.1 | 6, 7 | 872 | |||
XM_011511095.2 | XP_011509397.1 | 5, 6 | 642 | |||
XM_011511096.1 | XP_011509398.1 | 2, 3 | 252 | |||
XM_017003971.1 | XP_016859460.1 | 5, 6 | 818 | |||
XM_017003972.1 | XP_016859461.1 | 6, 7 | 922 | |||
XM_017003973.1 | XP_016859462.1 | 5, 6 | 828 |