Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCKAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207363.2 | NP_997246.2 | 8 | 927 | ||
NM_207481.3 | NP_997364.3 | 8 | 927 | |||
XM_005263659.3 | XP_005263716.1 | 7 | 1509 | |||
XM_005263660.3 | XP_005263717.1 | 7 | 1024 | |||
XM_011511097.2 | XP_011509399.1 | 9 | 1697 | |||
XM_011511098.2 | XP_011509400.1 | 9 | 1694 | |||
XM_011511099.2 | XP_011509401.1 | 8 | 1092 | |||
XM_011511100.2 | XP_011509402.1 | 8 | 1190 | |||
XM_011511101.1 | XP_011509403.1 | 6 | 600 | |||
XM_011511102.2 | XP_011509404.1 | 7 | 5026 | |||
XM_011511103.2 | XP_011509405.1 | 5 | 828 | |||
XM_011511104.1 | XP_011509406.1 | 5 | 479 | |||
XM_011511105.2 | XP_011509407.1 | 5 | 1270 | |||
XM_017003974.1 | XP_016859463.1 | 8 | 820 | |||
XM_017003975.1 | XP_016859464.1 | 7 | 1122 | |||
XM_017003976.1 | XP_016859465.1 | 9 | 1718 | |||
XM_017003977.1 | XP_016859466.1 | 6 | 1628 | |||
XM_017003978.1 | XP_016859467.1 | 2 | 1038 | |||
XM_017003979.1 | XP_016859468.1 | 8 | 1567 | |||
XM_017003980.1 | XP_016859469.1 | 5 | 828 | |||
GenBank mRNA | AK092189.1 | - | 6 | 602 | ||
AY946007.1 | - | 3 | 349 | |||
AY946008.1 | - | 3 | 349 |