Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMVK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323011.1 | NP_001309940.1 | 5 | 544 | ||
NM_001323012.1 | NP_001309941.1 | 5 | 551 | |||
NM_006556.3 | NP_006547.1 | 5 | 894 | |||
GenBank mRNA | AK130234.1 | - | 6 | 782 | ||
BC006089.1 | - | 5 | 877 | 3604167 | ||
BC007694.2 | - | 5 | 807 | 3954909 | ||
L77213.1 | - | 5 | 606 |