Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAET1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001788.3 | NP_001001788.2 | 5 | 1131 | ||
NR_130110.1 | 5 | 1031 | ||||
GenBank mRNA | AY172579.1 | - | 5 | 1057 | ||
AY172580.1 | - | 5 | 889 |