Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003951.2 | NP_001003951.1 | 10 | 1148 | ||
GenBank mRNA | AB186393.1 | - | 10 | 1148 |